Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prpf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002204.1 | NP_001002204.1 | 7 - 8 | 822 | 148 | |
XM_021467629.1 | XP_021323304.1 | 8 - 9 | 938 | 148 | ||
GenBank mRNA | BC074081.1 | - | 7 - 8 | 822 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |