Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pex11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103120.1 | NP_001096590.1 | 2 - 3 | 519 | 113 | |
GenBank mRNA | BC151942.1 | - | 2 - 3 | 519 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |