Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: galcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213111.1 | NP_998276.1 | 10 - 11 | 1168 | 94 | |
GenBank mRNA | BC049316.1 | - | 10 - 11 | 1168 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |