Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102384.1 | NP_001095854.1 | 7 | 3346 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Danio rerio Both primers and probe map within a single exon |