Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hs6st1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080203.1 | NP_001073672.1 | 2 | 651 | 74 | |
GenBank mRNA | DQ812994.1 | - | 2 | 651 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |