Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrps22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098735.1 | NP_001092205.1 | 1 - 2 | 131 | 155 | |
GenBank mRNA | BC142888.1 | - | 1 - 2 | 131 | 155 | |
BC150427.1 | - | 1 - 2 | 275 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |