Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105683.1 | NP_001099153.1 | 4 - 5 | 790 | 86 | |
XM_005157593.1 | XP_005157650.1 | 5 - 6 | 970 | 86 | ||
GenBank mRNA | AY260853.1 | - | 5 - 6 | 865 | 86 | |
BC053305.1 | - | 5 - 6 | 857 | 86 | ||
BC153631.1 | - | 4 - 5 | 790 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |