Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030263.1 | NP_001025434.1 | 7 - 8 | 840 | 74 | |
GenBank mRNA | BC098559.1 | - | 7 - 8 | 840 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |