Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccnd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045068.1 | NP_001038533.1 | 2 - 3 | 408 | 85 | |
GenBank mRNA | FJ648074.1 | - | 2 - 3 | 471 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |