Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083068.1 | NP_001076537.1 | 5 - 6 | 2190 | 92 | |
XM_005159560.3 | XP_005159617.1 | 5 - 6 | 2435 | 92 | ||
XM_009294101.1 | XP_009292376.1 | 4 - 5 | 1501 | 92 | ||
GenBank mRNA | BC161621.1 | - | 5 - 6 | 2427 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |