Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ghra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083578.1 | NP_001077047.1 | 5 - 6 | 807 | 90 | |
GenBank mRNA | BC134903.1 | - | 5 - 6 | 807 | 90 | |
EU649774.1 | - | 5 - 6 | 899 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |