Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhcga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089577.1 | NP_001083046.1 | 1 - 2 | 247 | 80 | |
GenBank mRNA | AB286867.1 | - | 1 - 2 | 272 | 80 | |
BC139524.1 | - | 1 - 2 | 247 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |