Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: opn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111164.1 | NP_001104634.1 | 1 - 2 | 331 | 63 | |
GenBank mRNA | EF043381.1 | - | 1 - 2 | 331 | 63 | |
KT008413.1 | - | 1 - 2 | 325 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Danio rerio Probe spans exons |