Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cadm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113552.1 | NP_001107024.1 | 3 - 4 | 294 | 63 | |
GenBank mRNA | CU638788.1 | - | 3 - 4 | 238 | 63 | |
EU182352.1 | - | 3 - 4 | 294 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |