Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200818.1 | NP_957112.1 | 5 - 6 | 584 | 55 | |
XM_005173309.3 | XP_005173366.1 | 5 - 6 | 656 | 55 | ||
XM_021481069.1 | XP_021336744.1 | 5 - 6 | 656 | 55 | ||
GenBank mRNA | BC059702.1 | - | 5 - 6 | 584 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |