Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgam1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201300.2 | NP_958457.2 | 2 - 3 | 545 | 114 | |
GenBank mRNA | BC054936.1 | - | 2 - 3 | 545 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |