Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001365553.1 | NP_001352482.1 | 3 - 4 | 376 | 111 | |
NM_212781.2 | NP_997946.1 | 3 - 4 | 376 | 111 | ||
GenBank mRNA | AY169369.1 | - | 3 - 4 | 336 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |