Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: got2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213379.1 | NP_998544.1 | 1 - 2 | 147 | 67 | |
GenBank mRNA | BC054684.1 | - | 1 - 2 | 147 | 67 | |
BC165803.1 | - | 1 - 2 | 118 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |