Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081769.1 | NP_001075238.1 | 25 - 26 | 3357 | 95 | |
XM_023651656.1 | XP_023507424.1 | 3357 | 95 | |||
XM_023651657.1 | XP_023507425.1 | 3743 | 95 | |||
GenBank mRNA | AY027883.1 | - | 25 - 26 | 3357 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Equus caballus Probe spans exons |