Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081903.2 | NP_001075372.1 | 3 - 4 | 379 | 79 | |
GenBank mRNA | AY536518.1 | - | 3 - 4 | 380 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Equus caballus Probe spans exons |