Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EQMHCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082507.2 | NP_001075976.1 | 4 - 5 | 901 | 64 | |
GenBank mRNA | CD536658.1 | - | 1 - 2 | 77 | 64 | |
DQ145613.1 | - | 3 - 4 | 728 | 64 | ||
DQ145615.1 | - | 3 - 4 | 733 | 64 | ||
X79893.1 | - | 901 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Equus caballus Probe spans exons |