Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082509.2 | NP_001075978.2 | 1 - 2 | 111 | 127 | |
GenBank mRNA | X98459.1 | - | 117 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Equus caballus Probe spans exons |