Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIBIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099443.1 | NP_001092913.1 | 1 | 549 | 64 | |
GenBank mRNA | AB302195.1 | - | 1 | 549 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Equus caballus Both primers and probe map within a single exon |