Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099769.2 | NP_001093239.2 | 2 - 3 | 374 | 99 | |
GenBank mRNA | AY005808.1 | - | 2 - 3 | 362 | 99 |
Gene Symbol: LOC111771935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023634817.1 | XP_023490585.1 | 480 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Equus caballus Inventoried Probe spans exons |