Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163885.2 | NP_001157357.1 | 1 - 2 | 123 | 81 | |
XM_005607002.3 | XP_005607059.1 | 170 | 81 | |||
XM_023626153.1 | XP_023481921.1 | 173 | 81 | |||
GenBank mRNA | AF143950.1 | - | 1 - 2 | 54 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Equus caballus Probe spans exons |