Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC28A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419716.5 | XP_419716.1 | 1 - 2 | 776 | 88 | |
GenBank mRNA | BX930415.2 | - | 1 - 2 | 289 | 88 | |
BX930751.1 | - | 1 - 2 | 234 | 88 | ||
BX935176.2 | - | 1 - 2 | 576 | 88 | ||
CR390708.1 | - | 1 - 2 | 97 | 88 | ||
CR523990.1 | - | 1 - 2 | 74 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |