Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015299728.2 | XP_015155214.1 | 3 - 4 | 470 | 74 | |
XM_015299730.2 | XP_015155216.1 | 3 - 4 | 471 | 74 | ||
GenBank mRNA | EF520346.1 | - | 1 - 2 | 49 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |