Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277360.1 | NP_001264289.1 | 2 - 3 | 323 | 148 | |
XM_025148143.1 | XP_025003911.1 | 3 - 4 | 239 | 148 | ||
XM_025148145.1 | XP_025003913.1 | 3 - 4 | 332 | 148 | ||
GenBank mRNA | BX931642.2 | - | 2 - 3 | 332 | 148 | |
BX935235.1 | - | 2 - 3 | 271 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |