Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277139.1 | NP_001264068.1 | 1 - 2 | 152 | 152 | |
GenBank mRNA | BX929849.2 | - | 1 - 2 | 45 | 152 | |
BX930843.2 | - | 1 - 2 | 61 | 152 | ||
BX935551.1 | - | 1 - 2 | 151 | 152 | ||
BX936086.2 | - | 1 - 2 | 120 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |