Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PATZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001232641.5 | XP_001232642.2 | 1 - 2 | 1454 | 62 | |
XM_004934371.3 | XP_004934428.1 | 1 - 2 | 1454 | 62 | ||
XM_004934372.3 | XP_004934429.1 | 1 - 2 | 1454 | 62 | ||
XM_004934373.3 | XP_004934430.1 | 1 - 2 | 1454 | 62 | ||
XM_004934374.3 | XP_004934431.1 | 1 - 2 | 1454 | 62 | ||
XM_004934376.3 | XP_004934433.1 | 1 - 2 | 1454 | 62 | ||
GenBank mRNA | CR385351.1 | - | 1 - 2 | 220 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |