Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001234098.5 | XP_001234099.3 | 6 - 7 | 3707 | 59 | |
GenBank mRNA | AJ851644.1 | - | 6 - 7 | 3192 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |