Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM21C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012593.2 | NP_001012611.2 | 30 - 31 | 124 | ||
XM_015288386.2 | XP_015143872.1 | 30 - 31 | 124 | |||
XM_015288389.2 | XP_015143875.1 | 29 - 30 | 124 | |||
XM_015288390.2 | XP_015143876.1 | 29 - 30 | 124 | |||
XM_025151585.1 | XP_025007353.1 | 30 - 31 | 124 | |||
XM_025151586.1 | XP_025007354.1 | 30 - 31 | 124 | |||
XM_025151587.1 | XP_025007355.1 | 30 - 31 | 124 | |||
XM_025151588.1 | XP_025007356.1 | 29 - 30 | 124 | |||
XM_025151589.1 | XP_025007357.1 | 29 - 30 | 124 | |||
GenBank mRNA | AJ851808.1 | - | 30 - 31 | 3871 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |