Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015279008.2 | XP_015134494.1 | 1 - 2 | 333 | 95 | |
XM_015279009.2 | XP_015134495.1 | 1 - 2 | 333 | 95 | ||
XM_025154079.1 | XP_025009847.1 | 1 - 2 | 333 | 95 | ||
GenBank mRNA | CR389722.1 | - | 1 - 2 | 81 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |