Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015279226.2 | XP_015134712.1 | 4 - 5 | 828 | 112 | |
XM_015279233.1 | XP_015134719.1 | 4 - 5 | 853 | 112 | ||
XM_025146857.1 | XP_025002625.1 | 4 - 5 | 1136 | 112 | ||
GenBank mRNA | CR353653.1 | - | 3 - 4 | 823 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |