Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025142431.1 | XP_024998199.1 | 4 - 5 | 834 | 108 | |
GenBank mRNA | BX929902.2 | - | 4 - 5 | 851 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |