Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277557.3 | NP_001264486.1 | 2 - 3 | 305 | 89 | |
XM_015290629.2 | XP_015146115.1 | 4 - 5 | 691 | 89 | ||
XM_025152828.1 | XP_025008596.1 | 3 - 4 | 472 | 89 | ||
XM_025152829.1 | XP_025008597.1 | 2 - 3 | 405 | 89 | ||
XM_025152830.1 | XP_025008598.1 | 2 - 3 | 580 | 89 | ||
XM_025152831.1 | XP_025008599.1 | 2 - 3 | 1006 | 89 | ||
GenBank mRNA | BX934812.1 | - | 2 - 3 | 305 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |