Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205225.2 | NP_990556.1 | 3 - 4 | 369 | 72 | |
GenBank mRNA | AF208228.1 | - | 2 - 3 | 347 | 72 | |
X81968.1 | - | 370 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |