Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012404.1 | NP_001012404.1 | 4 - 5 | 1000 | 104 | |
XM_015276322.2 | XP_015131808.1 | 4 - 5 | 3806 | 104 | ||
GenBank mRNA | AY885230.1 | - | 4 - 5 | 1000 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |