Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012861.1 | NP_001012879.1 | 6 - 7 | 659 | 97 | |
XM_015275856.2 | XP_015131342.1 | 6 - 7 | 1145 | 97 | ||
XM_015275857.2 | XP_015131343.1 | 6 - 7 | 726 | 97 | ||
XM_015275858.2 | XP_015131344.1 | 5 - 6 | 968 | 97 | ||
XM_015275859.2 | XP_015131345.1 | 5 - 6 | 944 | 97 | ||
GenBank mRNA | AJ719502.1 | - | 6 - 7 | 659 | 97 | |
CR338760.1 | - | 6 - 7 | 634 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |