Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX3X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030800.1 | NP_001025971.1 | 5 - 6 | 528 | 134 | |
XM_015301535.2 | XP_015157021.1 | 5 - 6 | 524 | 134 | ||
GenBank mRNA | AJ851409.1 | - | 5 - 6 | 528 | 134 | |
MH119574.1 | - | 5 - 6 | 415 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |