Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031117.1 | NP_001026288.1 | 3 - 4 | 375 | 111 | |
XM_015278244.2 | XP_015133730.1 | 4 - 5 | 698 | 111 | ||
GenBank mRNA | AJ719477.1 | - | 3 - 4 | 375 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |