Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAFAH1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030911.4 | NP_001026082.2 | 4 - 5 | 394 | 86 | |
XM_015297997.2 | XP_015153483.1 | 4 - 5 | 372 | 86 | ||
XM_025143186.1 | XP_024998954.1 | 4 - 5 | 318 | 86 | ||
GenBank mRNA | AJ719312.1 | - | 4 - 5 | 373 | 86 | |
CN232272.1 | - | 4 - 5 | 387 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |