Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030951.1 | NP_001026122.1 | 1 - 2 | 143 | 74 | |
GenBank mRNA | AJ719788.1 | - | 1 - 2 | 143 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |