Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031155.1 | NP_001026326.1 | 4 - 5 | 639 | 94 | |
XM_015285438.2 | XP_015140924.1 | 4 - 5 | 638 | 94 | ||
XM_015285439.2 | XP_015140925.1 | 4 - 5 | 879 | 94 | ||
GenBank mRNA | AJ720367.1 | - | 4 - 5 | 639 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |