Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030871.3 | NP_001026042.2 | 11 - 12 | 1309 | 68 | |
XM_015296863.2 | XP_015152349.1 | 12 - 13 | 4159 | 68 | ||
XM_015296864.2 | XP_015152350.1 | 12 - 13 | 4156 | 68 | ||
XM_015296865.2 | XP_015152351.1 | 12 - 13 | 4153 | 68 | ||
XM_015296866.2 | XP_015152352.1 | 11 - 12 | 4036 | 68 | ||
XM_025142506.1 | XP_024998274.1 | 12 - 13 | 4177 | 68 | ||
XM_025142508.1 | XP_024998276.1 | 11 - 12 | 4033 | 68 |
Set Membership: |
![]() ![]() ![]() |