Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BG-like antigen 1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099355.1 | NP_001092825.1 | 13 - 14 | 767 | 94 | |
XM_025155765.1 | XP_025011533.1 | 9 - 10 | 1181 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |