Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001305674.2 | NP_001292603.1 | 2 - 3 | 556 | 96 | |
NM_204335.3 | NP_989666.1 | 2 - 3 | 556 | 96 | ||
XM_015278173.2 | XP_015133659.1 | 5 - 6 | 3038 | 96 | ||
XM_015278174.2 | XP_015133660.1 | 4 - 5 | 729 | 96 | ||
XM_015278175.2 | XP_015133661.1 | 3 - 4 | 721 | 96 | ||
XM_015278176.2 | XP_015133662.1 | 3 - 4 | 634 | 96 | ||
XM_015278177.2 | XP_015133663.1 | 5 - 6 | 3037 | 96 | ||
XM_015278178.2 | XP_015133664.1 | 5 - 6 | 3037 | 96 | ||
XM_015278179.2 | XP_015133665.1 | 5 - 6 | 3036 | 96 | ||
GenBank mRNA | AF330009.1 | - | 2 - 3 | 442 | 96 | |
BU385029.1 | - | 2 - 3 | 556 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |