Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204473.1 | NP_989804.1 | 958 | 74 | ||
XM_015298880.2 | XP_015154366.1 | 939 | 74 | |||
XM_015298881.2 | XP_015154367.1 | 905 | 74 | |||
XM_015298882.2 | XP_015154368.1 | 752 | 74 | |||
GenBank mRNA | AY156089.1 | - | 958 | 74 | ||
BX935704.1 | - | 919 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus |