Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MATN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205072.1 | NP_990403.1 | 5 - 6 | 1092 | 74 | |
GenBank mRNA | AJ000055.1 | - | 5 - 6 | 1092 | 74 | |
CR389007.1 | - | 1 - 2 | 64 | 74 | ||
CR390328.1 | - | 1 - 2 | 58 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |