Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006541.1 | NP_001006541.1 | 7 - 8 | 849 | 144 | |
XM_015291021.2 | XP_015146507.1 | 7 - 8 | 827 | 144 | ||
GenBank mRNA | AJ719736.1 | - | 7 - 8 | 849 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |